Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ EDNRB HEK 293T DA Assay Kit

Catalog No. K1349
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1349 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1349 Supplier Thermo Scientific™ Supplier No. K1349

missing translation for 'validMainframeMsg'

GeneBLAzer™ EDNRB HEK 293T DA (Division Arrested) cells and EDNRB-NFAT-bla HEK 293T cells contain the human Endothelin type B (EDNRB) receptor stably integrated into the CellSensor™ NFAT-bla HEK 293T cell line.

This cell line is available as dividing cells by requesting a quote for K1224 from dividingcells@invitrogen.com

GeneBLAzer™ EDNRB HEK 293T DA (Division Arrested) cells and EDNRB-NFAT-bla HEK 293T cells contain the human Endothelin type B (EDNRB) receptor stably integrated into the CellSensor™ NFAT-bla HEK 293T cell line. CellSensor™ NFAT-bla HEK 293T cells (Cat. No. K1129) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells(NFAT) response element. Division Arrested (DA) cells are available in two configurations: an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates.

DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both EDNRB HEK 293T DA cells and EDNRB-NFAT-bla HEK 293T cells are functionally validated for Z'-factor and EC50 concentrations of ET-1 (Figure 1). In addition, EDNRB-NFAT-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

Endothelin Receptor Type B (ENDRB) mediates signaling of endothelin proteins that regulate several critical biological processes, including the development and function of blood vessels, the production of certain hormones, and the stimulation of cell growth and division. Defects in EDNRB mediated signaling have been implicated in a wide range of diseases, including cancer, hypertension and cardiovascular disorders.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000115.1
Assay Entry Cell-based beta lactamase reporter gene
Cell State Division-Arrested Cells
Content And Storage GeneBLAzer™ EDNRB HEK 293T DA Assay Kit
Each system contains sufficient division arrested (DA) cells & substrate to assay 1 x 384-well plate. (Other materials are required separately; please refer to the protocol).
• EDNRB HEK 293T DA cells
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg
• Solution D, 1 mL
• 2 protocols
• Certificate of Analysis

GeneBLAzer™ EDNRB HEK 293T DA cells
Each system contains sufficient division arrested (DA) cells to assay 10 x 384-well plates (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
• EDNRB HEK 293T DA cells
• 1 protocol
• Certificate of Analysis

GeneBLAzer™ EDNRB-NFAT-bla HEK 293T cells
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
• EDNRB-NFAT-bla HEK 293T cells
• 1 protocol
• Certificate of Analysis

Storage:
GeneBLAzer™ EDNRB HEK 293T DA Cells and EDNRB-NFAT-bla HEK 293T Cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt or thaw for immediate use.
Detection Method Fluorescence
Gene ID (Entrez) 1910
Gene Symbol EDNRB
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 1 plate
Reporter Gene BLA (Beta-Lactamase)
Target Entry EDNRB
Cell Line 293T (HEK)
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.