Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ GPR54 CHO-K1 DA Assay Kit

Catalog No. K1333
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1333 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1333 Supplier Thermo Scientific™ Supplier No. K1333

missing translation for 'validMainframeMsg'

GeneBLAzer™ GPR54 CHO-K1 DA (Division Arrested) cells and GPR54-NFAT-bla CHO-K1 cells contain the human KiSS1 receptor (GPR54) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.

This cell line is available as dividing cells by requesting a quote for K1181 from dividingcells@invitrogen.com

GeneBLAzer™ GPR54 CHO-K1 DA (Division Arrested) cells and GPR54-NFAT-bla CHO-K1 cells contain the human KiSS1 receptor (GPR54) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1078) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element. Division Arrested (DA) cells are available in two configurations: an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates.

DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both GPR54 CHO-K1 DA cells and GPR54-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of Metastin, (Figure 1). In addition, GPR54-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

The GPR54 receptor has been shown to be involved in tumor progression and metastasis, and in the development and functional regulation of the gonads.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_115940.1
Assay Entry Cell-based beta lactamase reporter gene
Cell State Division-Arrested Cells
Content And Storage GeneBLAzer™ GPR54 CHO-K1 DA Assay Kit
Each system contains sufficient division arrested (DA) cells & substrate to assay 1 x 384-well plate. (Other materials are required separately; please refer to the protocol).
• GPR54 CHO-K1 DA cells
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg
• Solution D, 1 mL
• 2 protocols
• Certificate of Analysis

GeneBLAzer™ GPR54 CHO-K1 DA cells
Each system contains sufficient division arrested (DA) cells to assay 10 x 384-well plates (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
• GPR54 CHO-K1 DA cells
• 1 protocol
• Certificate of Analysis

GeneBLAzer™ GPR54-NFAT-bla CHO-K1 cells
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
• GPR54-NFAT-bla CHO-K1 cells
• 1 protocol
• Certificate of Analysis

Storage:
GeneBLAzer™ GPR54 CHO-K1 DA Cells and GPR54-NFAT-bla CHO-K1 Cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt or thaw for immediate use.
Detection Method Fluorescence
Gene ID (Entrez) 84634
Gene Symbol GPR54
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 1 plate
Reporter Gene BLA (Beta-Lactamase)
Target Entry GPR54
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.