Learn More
Thermo Scientific™ GeneBLAzer™ Gqi5-NFAT-bla FreeStyle™ 293F Cells
Description
GeneBLAzer™ Gqi5-NFAT-bla FreeStyle™ 293F cells contain the chimeric G-protein Gqi5 stably integrated into the CellSensor™ NFAT-blaFreeStyle™ 293F cell line. CellSensor™ NFAT-bla FreeStyle™ 293F cells contain a beta-lactamase reporter gene under control of a stably integrated NFAT response element. Gqo5-NFAT-bla FreeStyle™ 293F is a parental cell line designed to be used for the construction of Gi/o-coupled GPCR beta-lactamase cellular assays. The Gi/o receptor of interest should be stably expressed in the cell line by expression from the selectable vector pcDNA™6.2-hygro-DEST (Cat. No. K1233). Gqi5-NFAT-bla FreeStyle™ 293F cells have been shown to respond to the agonist of a transiently expressed D2 receptor.
Specifications
Specifications
| Cell State | Dividing Cells |
| Content And Storage | Shipping: Dry Ice Storage: Liquid Nitrogen |
| Detection Method | Fluorescence |
| Gene Symbol | Gqi5 |
| Product Line | GeneBLAzer |
| Quantity | 2 x 106 Cells |
| Reporter Gene | BLA (Beta-Lactamase) |
| Type | Cell Line |
| Cell Line | FreeStyle 293-F Cells |
| System Type | GeneBLAzer™ |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.