Learn More
Thermo Scientific™ GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 Cells
Description
The GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line contains the promiscuous Gα protein, Gqo5, stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. NFAT-bla CHO-K1 cells (Cat. No. K1534) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element stably integrated into CHO-K1 cells. The Gqo5-NFAT-bla CHO-K1 Parental Cell line is designed for the construction of Gi/o coupled GPCR cell based assays.
Specifications
Specifications
| Cell State | Dividing Cells |
| Content And Storage | GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Detection Method | Fluorescence |
| Gene Symbol | Gα15 |
| Product Line | GeneBLAzer |
| Quantity | 2 x 106 Cells |
| Reporter Gene | BLA (Beta-Lactamase) |
| Cell Line | CHO-K1 |
| System Type | GeneBLAzer™ |
| Druggable Target | GPCR |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.