Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ HCRTR2-Gα15 CHO-K1 DA Assay Kit

Catalog No. K1369
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1369 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1369 Supplier Thermo Scientific™ Supplier No. K1369

missing translation for 'validMainframeMsg'

GeneBLAzer™ HCRTR2-Gα15 CHO-K1 DA(Division Arrested) cells and HCRTR2-Gα15-NFAT-bla CHO-K1 cells contain the human Hypocretin 2 receptor (HCRTR2) stably integrated into the GeneBLAzer™ Ga15-NFAT-bla CHO-K1 cell line.

This cell line is available as dividing cells by requesting a quote for Cat. No. K1246 from dividingcells@invitrogen.com

GeneBLAzer™ HCRTR2-Gα15 CHO-K1 DA(Division Arrested) cells and HCRTR2-Gα15-NFAT-bla CHO-K1 cells contain the human Hypocretin 2 receptor (HCRTR2) stably integrated into the GeneBLAzer™ Ga15-NFAT-bla CHO-K1 cell line. Gα15-NFAT-bla CHO-K1 cells (Cat. No. K1213) contain a beta-lactamase reporter gene under control of the nuclear factor of activated T-cells Response Element (NFAT), as well as the promiscuous G-protein, Ga15. Division Arrested (DA) cells are available in two configurations: an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates.

DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both HCRTR2-Gα15 CHO-K1 DA cells and HCRTR2-Gα15-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of Orexin B (Figure 1). In addition, HCRTR2-Gα15-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

HCRTR2 has high affinity for both orexin peptides. Expression of this receptor has been detected in the brain as well as adrenal gland, gastrointestinal tract, and pancreas. The signaling cascades used by orexin receptors appear to be varied and cell type dependant. In CHO cells, calcium release is seen after agonist treatment. In hypothalamic neurons, the frequency of action potentials is increased. Administering either orexin peptide into the bloodstream of rats caused an increase in feeding behavior. The orexin system is also thought to regulate neuroendocrine function and the sleep wake cycle. HCRTR2 has been directly implicated in narcolepsy in animal studies.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_001526.2
Assay Entry Cell-based beta lactamase reporter gene
Cell State Division-Arrested Cells
Content And Storage GeneBLAzer™ HCRTR2-Gα15 CHO-K1 DA Assay Kit
Each system contains sufficient division arrested (DA) cells & substrate to assay 1 x 384-well plate. (Other materials are required separately; please refer to the protocol).
• HCRTR2-Gα15 CHO-K1 DA cells
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg
• Solution D, 1 mL
• 2 protocols
• Certificate of Analysis

GeneBLAzer™ HCRTR2-Gα15 CHO-K1 DA cells
Each system contains sufficient division arrested (DA) cells to assay 10 x 384-well plates (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
• HCRTR2-Gα15 CHO-K1 DA cells
• 1 protocol
• Certificate of Analysis

GeneBLAzer™ HCRTR2-Gα15-NFAT-bla CHO-K1 cells
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
• HCRTR2-Gα15-NFAT-bla CHO-K1 cells
• 1 protocol
• Certificate of Analysis

Storage
GeneBLAzer™ HCRTR2-Gα15 CHO-K1 DA Cells and HCRTR2-Gα15-NFAT-bla CHO-K1 Cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt or thaw for immediate use.
Detection Method Fluorescence
Gene ID (Entrez) 3062
Gene Symbol HCRTR2
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 1 plate
Reporter Gene BLA (Beta-Lactamase)
Target Entry HCRTR2
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.