Learn More
Thermo Scientific™ GeneBLAzer™ HTR2C-NFAT-bla CHO-K1 Cells
Description
GeneBLAzer™ HTR2C-NFAT-bla CHO-K1 cells contain the human 5-hydroxytrptamine (serotonin) receptor 2C (HTR2C), (Accession No. NM_000868.1) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1078) contain a beta-lactamase reporter gene under control of the NFAT Response Element. The GeneBLAzer™ HTR2C-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of 5-Hydroxytrptamine. In addition, GeneBLAzer™ HTR2C-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions.
Specifications
Specifications
| Accession Number | NM_000868.1 |
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | GeneBLAzer™ cell lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Detection Method | Fluorescence |
| For Use With (Application) | FRET |
| Human Protein Class | G-Protein Coupled Receptor (GPCR) |
| Population Status | Single Cell Clone Origin |
| Product Line | GeneBLAzer |
| Quantity | 8 x 106 cells/vial |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.