Learn More
Thermo Scientific™ GeneBLAzer™ Jurkat Control Kit
Catalog No. K1206
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
K1206 1 kit
1 options
Catalog No. K1206 Supplier Thermo Scientific™ Supplier No. K1206
The GeneBLAzer™ Jurkat Control Kit contains constitutively expressing (CMV-bla) and wildtype Jurkat cells to provide positive and negative controls, respectively, for evaluating beta-lactamase activity (Figure 1).
The GeneBLAzer™ Jurkat Control Kit contains constitutively expressing (CMV-bla) and wildtype Jurkat cells to provide positive and negative controls, respectively, for evaluating beta-lactamase activity (Figure 1).
TRUSTED_SUSTAINABILITY
Specifications
Cell State Dividing Cells
Content And Storage The GeneBLAzer™ Jurkat Control Kit contains 2 x 1 mL vials of cell s(∼5 x 106 cells/mL) and is shipped on dry ice. Store in liquid nitrogen upon receipt.
Detection Method Fluorescence
Product Line GeneBLAzer
Quantity 1 kit
Cell Line Jurkat
System Type GeneBLAzer™
Druggable Target Signaling Pathway
For Research Use Only. Not for use in diagnostic procedures.