Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ M1 CHO-K1 DA Assay Kit

Catalog No. K1365
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1365 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1365 Supplier Thermo Scientific™ Supplier No. K1365

GeneBLAzer™ M1 CHO-K1 DA(Division Arrested) cells and M1-NFAT-bla CHO-K1 cells contain the human Acetylcholine (muscarinic) subtype 1 receptor (M1) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.

This cell line is available as dividing cells by requesting a quote for K1230 from dividingcells@invitrogen.com

GeneBLAzer™ M1 CHO-K1 DA(Division Arrested) cells and M1-NFAT-bla CHO-K1 cells contain the human Acetylcholine (muscarinic) subtype 1 receptor (M1) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1078) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element. Division Arrested (DA) cells are available in two configurations: an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates.

DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both M1 CHO-K1 DA cells and M1-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of carbachol, (Figure 1). In addition, M1-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

The Musacarinic 1(M1) Receptor is localized principally in the cortical and hippocampal regions of the brain and has been implicated in various neurological conditions including depression, anxiety, and sleep disorders.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000738
Assay Entry Cell-based beta lactamase reporter gene
Cell State Division-Arrested Cells
Content And Storage GeneBLAzer™ M1 CHO-K1 DA Assay Kit
Each system contains sufficient division arrested (DA) cells & substrate to assay 1 x 384-well plate. (Other materials are required separately; please refer to the protocol).
• M1 CHO-K1 DA cells
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg
• Solution D, 1 mL
• 2 protocols
• Certificate of Analysis

GeneBLAzer™ M1 CHO-K1 DA cells
Each system contains sufficient division arrested (DA) cells to assay 10 x 384-well plates (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol)..
• M1 CHO-K1 DA cells
• 1 protocol
• Certificate of Analysis

GeneBLAzer™ M1-NFAT-bla CHO-K1 cells
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
• M1-NFAT-bla CHO-K1 cells
• 1 protocol
• Certificate of Analysis

Storage:
GeneBLAzer™ M1 CHO-K1 DA Cells and M3-NFAT-bla CHO-K1 Cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt or thaw for immediate use.
Detection Method Fluorescence
Gene ID (Entrez) 1128
Gene Symbol M1
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 1 plate
Reporter Gene BLA (Beta-Lactamase)
Target Entry M1
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.