Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ M4-Gqo5-NFAT-bla CHO-K1 Cells

Catalog No. K1741
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 106 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1741 2 x 106 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1741 Supplier Thermo Scientific™ Supplier No. K1741
Only null left
Add to Cart
Add to Cart
Request a Quote

GeneBLAzer™ M4 Gqo5-NFAT-bla CHO-K1 cells contain the human Acetylcholine (muscarinic) subtype 4 receptor (M4) stably integrated into the GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line.

GeneBLAzer™ M4 Gqo5-NFAT-bla CHO-K1 cells contain the human Acetylcholine (muscarinic) subtype 4 receptor (M4) stably integrated into the GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line. GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cells (Cat. No. K1536) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element and the Chimeric G protein, Gqo5.

The GeneBLAzer™ M4 Gqo5-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of Cabachol. In addition, GeneBLAzer™ M4 Gqo5-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

The clinical implications of the M4 receptor are unknown, however studies with knockout mice suggest that M4 may have implications in dopaminergic function, karatinocyte migration and wound healing, anxiolysis, and analgesia. In addition, M4 may be important in memory circuits and have implications in Alzheimer's Disease. Additional information on the muscarinic receptors can be found in literature.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000741
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ M4 Gqo5 CHO-K1 DA cells
• GeneBLAzer™ M4 Gqo5 CHO-K1 DA Assay Protocol
• Certificate of Analysis
• Solution D
• LiveBLAzer™-FRET B/G Loading Protocol

The GeneBLAzer™ M4-Gqo5-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol M4
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry M4
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.