Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ OPRL1-Gqi5-NFAT-bla FreeStyle™ 293F Cells

Catalog No. K1714
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 106 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1714 2 x 106 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1714 Supplier Thermo Scientific™ Supplier No. K1714
Only null left
Add to Cart
Add to Cart
Request a Quote

The GeneBLAzer™ OPRL1-Gqi5-NFAT-bla FreeStyle™ 293F Cell Line contains the human opioid/opiate receptor-like-1 receptor (OPRL1) stably integrated into the GeneBLAzer™ Gqi5-NFAT-bla FreeStyle™ 293F Cell Line.

The GeneBLAzer™ OPRL1-Gqi5-NFAT-bla FreeStyle™ 293F Cell Line contains the human opioid/opiate receptor-like-1 receptor (OPRL1) stably integrated into the GeneBLAzer™ Gqi5-NFAT-bla FreeStyle™ 293F Cell Line. GeneBLAzer™ Gqi5-NFAT-bla FreeStyle™ 293F cells (Cat. No. K1539) contain the chimeric Gqi5 G protein and the beta-lactamase reporter gene under control of the nuclear factor of activated T cells (NFAT) response element.

OPRL1-Gqi5-NFAT-bla FreeStyle™ 293F cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time, and functionally validated for Z-factor and EC50 concentrations of orphanin FQ/nociceptin. These data are found in the Validation & Assay Performance Summary.

Orphanin FQ/nociceptin and OPRL1 have been implicated in various physiological functions. The most defined role is their involvement in the mediation of pain in the central nervous system and their possible roles in opiate tolerance, opiate dependence and withdrawal, and the ability to cope with stress and anxiety. Through the use of antagonists, antisense oligos, and transgenic knockout mutants, OPRL1 has been implicated in additional systems including learning, memory, and reproduction.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000913
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ OPRL1-Gqi5-NFAT-bla Freestyle 293F cells
• GeneBLAzer™ OPRL1-Gqi5-NFAT-bla Freestyle 293F Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ OPRL1-Gqi5-NFAT-bla Freestyle 293F cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol OPRL1
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry OPRL1
Type Cell Line
Cell Line FreeStyle 293-F Cells
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.