Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ PPAR gamma DA Assay Kit

Catalog No. K1419
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1419 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1419 Supplier Thermo Scientific™ Supplier No. K1419

GeneBLAzer™PPAR gamma DA (Division Arrested) cells and PPAR gamma-UAS-bla HEK 293H cells contain the ligand-binding domain (LBD) of the human Peroxisome Proliferator-Activated Receptor-gamma (PPAR gamma) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293H cell line.

GeneBLAzer™PPAR gamma DA (Division Arrested) cells and PPAR gamma-UAS-bla HEK 293H cells contain the ligand-binding domain (LBD) of the human Peroxisome Proliferator-Activated Receptor-gamma (PPAR gamma) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293H cell line. GeneBLAzer™UAS-bla HEK 293H cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-PPAR gamma (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both PPAR gamma DA cells and PPAR gamma-UAS-bla HEK 293H cells are functionally validated for Z' and EC50 concentrations of Rosiglitazone. In addition, PPAR gamma-UAS-bla HEK 293H cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, and stimulation time.

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Cell-based beta lactamase reporter gene
Cell State Division-Arrested Cells
Content And Storage Contains sufficient division-arrested cells and substrate to assay 1 x 384-well plate:
• PPAR gamma 293H DA cells (K1419A)
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen)
Detection Method Fluorescence
Gene ID (Entrez) 5468
Gene Symbol PPARγ
Primary Agonist Calcitriol
Product Line GeneBLAzer
Quantity 1 plate
Reporter Gene BLA (Beta-Lactamase)
Target Entry PPARG, PPAR gamma, NR1C3
Type Division-Arrested Cells Assay Kit
Cell Line 293H
System Type GeneBLAzer™
Druggable Target Nuclear Receptors
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.