Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ GeneMapper™ ID-X Software v1.6, full installation

Catalog No. A39975
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 license
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A39975 1 license
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A39975 Supplier Applied Biosystems™ Supplier No. A39975
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GeneMapper ID-X Software is an automated genotyping solution for forensic casework and databasing data analysis, as well as for paternity data analysis.

  • GeneMapper ID-X Software is an automated genotyping solution for forensic casework and databasing data analysis, as well as for paternity data analysis
  • It supports a multi-user database configuration and provides a set of streamlined data analysis and sample review tools
  • It also offers a comprehensive quality value system that is designed to be optimized and validated for use in both expert system and traditional data analysis workflows
  • GeneMapper ID-X Software supports genotyping applications on the Applied Biosystems 310, 3100, 3130, 3130xl, 3500, 3500xl, 3730, and SeqStudio genetic analyzers
  • The release notes for this version provide information that should allow laboratories to decide whether internal validation testing is necessary for its system to comply with laboratory guidelines or regulations

GeneMapper ID-X Software v1.6 features include:

  • Supported operating systems: Windows™ 7 Professional, 64-bit (Service Pack 1); Windows 10 Enterprise, 64-bit; Windows 10 Enterprise 2016 LTSB, 2019 LTSC
  • Support for Java version 8
  • Latest oracle security patches
  • Support for CODIS 3.2 Revision 16 and CODIS 3.3 specifications
  • Internationalized with local language support for English, Russian, and Chinese
  • Enhanced command line interface functionality
  • Enhanced user interfaces for better usability
  • Enhanced printing
  • View Table by Marker functionality in Report Manager and export as PDF
  • Enhanced profile comparison
  • Enhanced Security and Audit Module

Overview of GeneMapper ID-X Software:

  • Powerful, easy-to-use data analysis tool designed to increase lab productivity
  • Comprehensive Expert System capability enables analysis of forensic databasing samples delivered by automated data assessment functionality and efficient manual review tools
  • Complete Expert Assistant solution for analysis of forensic casework samples combines an efficient suite of manual review features with an integrated mixture analysis tool
  • Quality control functionality allows rapid allele match comparisons and concordance searches within a data set
  • Extensive security, auditing, and e-signature capabilities help protect data integrity and control technical records
  • Multi-user database configuration stores your projects in a central location and facilitates information exchange
  • Command Line Interface offers the ability to access GeneMapper ID-X Software automatically (without human intervention)
  • It also helps streamline its integration with LIMS installations and other applications in an existing laboratory infrastructure
TRUSTED_SUSTAINABILITY

Specifications

Product Line GeneMapper
Software Type License
Content And Storage Store at room temperature.
Product Type GeneMapper ID-X Software V1.6 Full Installation
Quantity 1 license
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.