Learn More
Applied Biosystems™ GeneMapper™ Software v6.1, full installation
Catalog No. A53547
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 license
Catalog No. Quantity
A53547 1 license
1 options
Catalog No. A53547 Supplier Applied Biosystems™ Supplier No. A53547
This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or visit www.fishersci.com/quantstudio for more information and to contact a Life Science Specialist.
GeneMapper Software is a flexible genotyping software package that provides DNA sizing and quality allele calls for all Thermo Fisher Scientific electrophoresis-based genotyping systems.
  • GeneMapper Software is a flexible genotyping software package that provides DNA sizing and quality allele calls for all Thermo Fisher Scientific electrophoresis-based genotyping systems
  • This software specializes in multi-application functionality, including amplified fragment length polymorphism (AFLP), loss of heterozygosity (LOH), microsatellite and SNP genotyping analysis
  • GeneMapper Software can help users increase data processing efficiency with remote auto-analysis and command line operation, and allows for multiuser, client-server deployment
  • The software uses Process Quality Values (PQVs) for automated identification that reduces data review time for high throughput genotyping
  • In addition, the security and audit features help users meet 21 CFR 11 requirements
  • GeneMapper Software has not undergone specific developmental validation for human identification applications
  • More than five clients simultaneously accessing the database may slow the performance of GeneMapper Software.
Features include:
  • Installation on the data collection computer or a non-instrument computer
  • Support of networked multiple clients when installed on a non-instrument computer
  • An Oracle database with project-related data storage that ensures faster installation times and flexible archive capabilities
  • Customizable project auto-saving frequency
  • Defining of linearity range in the analysis methods for more reproducible, higher quality sizing of larger fragments (>800 bps)
  • Analysis of up to 8-dye data
Recommended Computer Configuration
The software is tested with the following specifications:
  • Dell OptiPlex XE3 Windows™ 10 Enterprise LTSC operating system
  • Dell 3551 Windows™ 10 Enterprise LTSC operating system
  • CPU: Core i5-6440HQ/3.0GHz and CoreTM i5-10400H CPU @ 2.60 GHz
  • Memory: 16GB
  • Disk: Free space: 500MB on the boot drive, 7 GB on the drive on which the GeneMapper™ Software is installed
TRUSTED_SUSTAINABILITY
Specifications
Product Line GeneMapper
Software Type CE Fragment Analysis
Content And Storage The software is provided in the form of a USB stick.
Data Entry CE Fragment Analysis (.fsa files)
License Full
Operating System Windows 10
Quantity 1 license
How can I set up auto-analysis for data in GeneMapper with the 3500 Data Collection Software version 3.x?
Beginning with 3500 Data Collection Software version 3, the 3500 Data Collection Software does not support automatic secondary analysis. This is also indicated in the software release notes. To perform secondary data analysis, you will need to manually import the .fsa file into the GeneMapper software. For optimal performance, we recommend that you install the GeneMapper Software on a stand-alone computer that is not part of the 3500 Genetic Analyzer system.
Can I install GeneMapper Software v6.1, full installation and GeneMapper Software v6.1, client installation on the same computer?
No, GeneMapper Full Software and GeneMapper Client Software must be installed on different computers.
How many user accounts can be connected to the Oracle database embedded in GeneMapper Software v6.1, full installation at any time?
Five user accounts can be connected to the Oracle database embedded in GeneMapper Software v6.1, full installation at any time.
We just upgraded to GeneMapper Software v6.1, and I'm wondering if my old data files will work on it.
GeneMapper Software v6.1 is compatible with the following files:
- 3130 series, Data Collection Software v3.0, 3.1, 4
- 3730 series, Data Collection Software v3.x, 4.x
- 3500 series, Data Collection Software v1.x, 2, 3.x
- SeqStudio Data Collection Software v1.x
- SeqStudio Flex Series Genetic Analyzer, Data Collection Software v1.x
Where can I download the Release Notes for GeneMapper Software v6.1, full installation?
Please download this PDF (https://downloads.thermofisher.com/Instrument_Software/Genemapper_6.1_Full/100107864_GeneMapper-Full_v6.1_Release_Notes.pdf).
Can I install GeneMapper Software v6.1 on Windows 7 operating system?
No, GeneMapper Software v6.1 supports only Windows 10 Professional/Enterprise operating system.
Which instrument data does GeneMapper Software v6.1 support?
GeneMapper Software v6.1 supports data generated from the following Applied Biosystems instruments:
- SeqStudio Flex Series Genetic Analyzers
- SeqStudio Genetic Analyzer
- 3500 Genetic Analyzer
- 3500xL Genetic Analyzer
- 3130 Genetic Analyzer
- 3130xl Genetic Analyzer
- 3730 DNA Analyzer
- 3730xl DNA Analyzer
- 310 Genetic Analyzer
Do you have a manual of install/uninstall procedures for GeneMapper Software v6?
Please download this PDF (https://downloads.thermofisher.com/Instrument_Software/CE/GeneMapper/Full_v6.0/100079198_MNL_GENEMAPPER_V_6_INSTALLATION.pdf) and follow the directions for install/uninstall GeneMapper Software v6.
What information should I back up when using GeneMapper Software?
It is useful to back up data in order to protect yourself from data loss, clearing out the Oracle database to keep it from filling up, and/or for sending data to Technical Support for troubleshooting.
In GeneMapper Software, there are 3 locations where you will need to back up data:
1. Panel Manager - contains information on your Panels, Markers, and Bins.
2. GeneMapper Manager - contains information pertaining to the Project itself - Projects, Analysis Methods, Size Standard Definitions, Table Settings, Plot Settings, Matrices (ABI PRISM 310 Genetic Analyzer Only), Cluster Plot Settings, SNP Sets, and Report Settings.
3. Data Files - sample data (.fsa files) that were used in the Project.
Note: Older versions of GeneMapper Software contained an Archive function that would back up the entire database at once. However, the archive could only be reimported into the same instance of GeneMapper Software. If you had a problem that required reinstallation of GeneMapper Software, it would be considered a new instance of the software and the Archive could not be imported into it.
I tried to launch GeneMapper Software and am getting a "Cannot connect to the Database" error message. The software is installed in a Client/Server configuration (Client and Server are on different computers). What should I do?
Please download this PDF (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-troubleshooting-client-install.pdf) for directions on how to resolve this error.
I tried to launch GeneMapper Software and am getting a "Cannot connect to the Database" error message. The software is installed on a stand-alone configuration, where the computer is not connected to the instrument. What should I do?
Please download this PDF (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-troubleshooting-full-install.pdf) for directions on how to resolve this error.
I tried to launch GeneMapper Software and am getting a "Cannot connect to the Database" error message. The software is installed on the instrument computer, where my computer is connected to the instrument (Client and Server on same computer). What should I do?
Please download this PDF (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-troubleshooting-full-install-instrument.pdf) for directions on how to resolve this error.
My Oracle database became corrupted and I have to reinstall the GeneMapper Software. However, when I try to uninstall using Add/Remove programs, it won't work. How do I manually uninstall GeneMapper Software?
Before uninstalling, make sure you have your GeneMapper CD/DVD and your registration code for reinstallation. Follow the uninstall instructions for your version of GeneMapper Software. Before attempting to reinstall, verify that you are logged in locally as the Administrator, that your antivirus Software is either off or that you have suspended activity during installation, and that your TCP/IP is installed.
For Research Use Only. Not for use in diagnostic procedures.