Learn More
Amsbio LLC GenePORTER Transfection reagent
Supplier:  Amsbio LLC AMS.T201075
Encompass
The GenePORTER Reagent is the original transfection lipid developed by Genlantis containing a unique formulation of the neutral lipid dioleoyl phosphatidylethanolamine (DOPE) and using the proprietary and innovative Direct Hydrophilic Conjugation (DHC) technology. The GenePORTER reagent is optimized for serum-free transfection conditions in a wide variety of commonly used cells. The GenePORTER reagent is especially effective in Jurkat cells and gives the highest transfection efficiencies relative to all other commercial reagents.
Catalog No. 50-163-2237