Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega GenePrint 24 System, 100 reactions

Catalog No. PRB1870 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
No. of Reactions:
100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
PRB1870 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRB1870 Supplier Promega Supplier No. B1870
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The GenePrint 24 System is a 24-locus multiplex system designed to generate a multi-locus human DNA profile from a variety of human-derived biological sources.

  • Confidence for Mixed Sample Analysis
  • Track and confirm tissue specimen provenance and cell lineage
  • Detect contaminants, including extraneous tissue or mixed cell populations
  • Resolve complex mixtures from related individuals or multiple sources

Specifications

Content And Storage -30°C to +10°C
No. of Reactions 100
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.