Learn More
Alpco Ltd GeneProof Treponema pallidum PCR Kit
SDP
Supplier:  Alpco Ltd 60TP002100
Encompass_Preferred
The GeneProof Treponema pallidum PCR Kit targets the highly conservative polA gene for DNA polymerase I. For Research Use Only in North America. This kit is sufficient for 100 tests.
Catalog No. NC2284581
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.