Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ General Assay Microplate Lids
BIOPHARMAGSA_VAGREENER_CHOICE

Catalog No. 07200598 Shop All Corning Products
Change view
Click to view available options
Plate Compatibility:
96-well Microplates Only (not 2 mL block)
96/384-well, Half Area Plate
All Microplates
No. per Case:
100/Cs.
50/Cs.
Material:
Polystyrene
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plate Compatibility No. per Case Material
07-200-598 96-well Microplates Only (not 2 mL block) 50/Cs. Polystyrene
07-200-703 96/384-well, Half Area Plate 50/Cs. Polystyrene
07-200-694 All Microplates 100/Cs. Polystyrene
07-200-597 96-well Microplates Only (not 2 mL block) 100/Cs. Polystyrene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 07-200-598 Supplier Corning™ Supplier No. 3931
Add to Cart
Add to Cart
This item is not returnable. View return policy

Manufactured with rigid polystyrene for optimum visibility and strength. Corning™ General Assay Microplate Lids are available in a variety of styles and types.

  • All lids are made of rigid polystyrene except where indicated.
  • All lids have a corner notch on the A1 corner (except where indicated) to correspond to the corner notches found on all Corning™ microplates.
  • The universal lid without a corner notch (Cat. No. 3098) does not need to be oriented in any particular direction to be placed on Corning microplates. The lid also has a shorter skirt than standard lids.
  • The black universal lid (Cat. No. 3935) is suitable for fluorescent and other light-sensitive assays.
  • The DMSO-resistant cyclic olefin copolymer (COC) lid (Cat. No. 3085) is tinted amber in color for light-sensitive assays and is 100% DMSO-resistant.
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    TRUSTED_SUSTAINABILITY

    Specifications

    Description Low evaporation lid with corner notch and condensation rings
    Material Polystyrene
    Sterility Sterile
    Condensation Rings Yes
    No. per Case 50/Cs.
    No. per Pack 25/Pk.
    Product Type Lid
    Plate Compatibility 96-well Microplates Only (not 2 mL block)
    Cut-off Corners Corner Notch
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.