Learn More
General Biology Slide Set D
SCI_ED
Catalog No. S750 Shop All Johannes Lieder GmbH & Co. KG Products
Change view
Click to view available options
Quantity:
50
Catalog No. Quantity
S750 50
1 options
This item is not returnable. View return policy
Catalog No. S750 Supplier Johannes Lieder GmbH & Co. KG Supplier No. 750
This item is not returnable. View return policy
Set D offers prepared slides of animal and plant structures in whole mount (w.m.), longitudinal section (l.s.) and cross-section (c.s.) preparations. Also available in CD-ROM format (see S750CD, below).
Histology of Humans and Mammals
  • Ciliated epithelium, t.s., fallopian tube of pig
  • Tendon, l.s., white fibrous tissue, fibers/cells
  • Heart muscle, t.s. and l.s., human, branched fibers with central nuclei and intercalated discs
  • Lymph gland, c.s., pig, lymphoid tissue
  • Esophagus, c.s., cat, stratified squamous epithelium
  • Stomach, c.s., cat, fundic with gastric glands
  • Large intestine, c.s., stained for mucous cells
  • Pancreas, pig, showing islets of Langerhans
  • Thyroid gland, t.s., pig, glandular epithelium and colloid
  • Adrenal gland, t.s., cat, through cortex and medulla
  • Sperm, smear, bull spermatozoa
  • Motor Nerve Cells, smear, spinal cord of cow
  • Cerebrum, t.s., human, cortex with pyramidal cells
  • Skin, human, palm, cornified epidermis, sweat glands
Zoology
  • Fasciola hepatica, w.m., beef liver fluke
  • Taenia, w.m., tapeworm mature proglottids
  • Culex pipiens, w.m. (F), mosquito head and mouth parts
  • Culex pipiens, w.m., male head and mouth parts
  • Cimex lectularis, w.m., bed bug, adult
Cytology and Genetics
  • Mitochondria, thin section, liver or kidney
  • Golgi apparatus, t.s., through spinal ganglion
  • Chloroplasts, in leaf Elodea or Mnium
  • Aleurone grains, sec. of Ricinus endosperm
  • Storage, c.s., liver or kidney vital stained with trypan-blue for storage in epithelial cells
  • DNA in cell nuclei, Feulgen stained
  • DNA and RNA, fixed and stained with methyl green to show DNA and RNA in different colors
  • Giant chromosomes, from Chirionomus salivary gland, individual genes and puffs can be seen
  • Human chromosomes, spread in metaphase for counting
  • Meiotic and mitotic stages, c.s., crayfish testis, with nuclear spindles
  • Maturation divisions, in ova of Ascaris megalocephala, iron-hematoxylin stained
  • Cleavage stages, in ova of Ascaris meg., iron-hematoxylin stained
Bacteria and Diseased Human Organs
  • Escherichia coli, bacteria from colon, Gram stained
  • Eberthella typhi, typhoid fever, Gram stained, smear
  • Tuberculosis, c.s., lung with tubercles in tissue
  • Smoker's lung, c.s., human smoker's lung
  • Liver cirrhosis, c.s., man, alcohol abuse
  • Arteriosclerosis, c.s., human coronary artery with sclerotic wall
  • Metastatic carcinoma (cancer), human liver
Embryology
  • Sea urchin development, 2/4/8 cell stages
  • Sea urchin development, morula, blastula, and gastrula
  • Frog embryology, c.s., through blastula/blastocoel
  • Frog embryology, c.s., larva, through organs
Ecology and the Environment
  • Leaf, needle of fir, two types, healthy and acid-rain damaged
  • Leaf of beech, c.s., two types, healthy and acid-rain damaged
  • Bacteria from wastewater, smear
Botany
  • Nostoc, filamentous colonies in gelatinous sheaths
  • Desmids, strewn, mixed species
  • Spahgnum, w.m., peat moss, leaf showing chlorophyll-bearing hyaline cells
  • Triticum, c.s., wheat stem with central pith and circular arrangement of bundles
  • Salvia, c.s., sage square stem, angular collenchyma
Specifications
Type General Biological Set
Quantity 50