Learn More
Description
- Human Serum Based (Lyophilized): Chemistry Control (Level 1)(23-666-187), Chemistry Control (Level II)(23-666-188), and Chemistry Control (Level 1 and II)(NC9896982)
- Hemoglobin Control Set (23-666-307) contains Human Erythrocytes (Lyophilized). Contains three levels of hemoglobin.
- Lipid Control Set (23-666-359) is Human Serum Based (Lyophilized). Expected Values: The lipid values were determined by repetitive assays of the listed methods. Values listed are targets only.
- Measurements using other reagents and instrument systems may give different results
- Listed ranges obtained from those suggested by the Health Care Finance Administration as acceptable performance criteria for deviation from a target value
- Due to method differences and matrix interactions, the values listed may not always be reproduced by other analytical systems
- Each laboratory should establish its own precision parameters
Specifications
Specifications
| Certifications/Compliance | CE |
| Content And Storage | 2°C to 8°C |
| Type | Ammonia or Alcohol Control Set |
| Intended Use | In-vitro diagnostic use, monitoring the accuracy and precision of various Ammonia and/or Ethanol assay methods and to validate quantitation of patient samples |
| Quantity | 2 x 5 mL |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.