Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
- Home
- Products
- Research and Clinical Diagnostic Tests and Controls
- Chemistry Controls, Calibrators and Reagents
- Clinical Chemistry Standards
- Pointe Scientific General Chemistry Standards
Pointe Scientific General Chemistry Standards
Catalog No. 23666277 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 2 mL
1 x 3 mL
1 x 4 mL
1 x 5 mL
1 x 15 mL
1 x 30 mL
Type:
Alcohol Standard
Ammonia Standard
BUN Standard
Calcium Standard
Carbon Dioxide Standard
Cholesterol - HDL Standard
Cholesterol Standard
Creatinine Standard
Glucose Standard
Hemoglobin Standard
Iron Standard
Lactate Standard
Lipase Standard Reagent Set
Magnesium Standard Reagent Set
Microprotein Standard Set
Total Protein Standard Set
Triglyceride Standard
Uric Acid Standard
Concentration:
0.001 to 0.002 g/dL
0.002 g/dL
0.005 g/dL
0.01 g/dL
0.02 g/dL
0.05 g/dL
0.1 g/dL
0.2 g/dL
100 μm/L
12 g/dL
30 mM/L
5 mM/L
500 μg/dL
500 μm/L
600 U/L
8 g/dL
19 product options available for selection
| Catalog No. | Quantity | Type | Concentration |
|---|---|---|---|
| 23-666-277 | 1 x 15 mL | Glucose Standard | 0.1 g/dL |
| 23-666-356 | 1 x 3 mL | Lipase Standard Reagent Set | 600 U/L |
| 23-666-424 | 1 x 15 mL | Uric Acid Standard | 0.005 g/dL |
| 23-666-199 | 1 x 15 mL | Cholesterol - HDL Standard | 0.05 g/dL |
| 23-666-341 | 1 x 15 mL | Lactate Standard | 5 mM/L |
| 23-666-360 | 1 x 15 mL | Magnesium Standard Reagent Set | 0.001 to 0.002 g/dL |
| 23-666-101 | 1 x 4 mL | Ammonia Standard | 500 μm/L |
| 23-666-182 | 1 x 15 mL | Carbon Dioxide Standard | 30 mM/L |
| 23-666-312 | 1 x 30 mL | Iron Standard | 500 μg/dL |
| 23-666-167 | 1 x 15 mL | Calcium Standard | 0.01 g/dL |
| 23-666-100 | 1 x 4 mL | Ammonia Standard | 100 μm/L |
| 23-666-071 | 1 x 5 mL | Alcohol Standard | 0.1 g/dL |
| 23-666-373 | 1 x 15 mL | Microprotein Standard Set | 0.05 g/dL |
| 23-666-153 | 1 x 15 mL | BUN Standard | 0.02 g/dL |
| 23-666-235 | 1 x 15 mL | Creatinine Standard | 0.002 g/dL |
| 23-666-422 | 1 x 15 mL | Triglyceride Standard | 0.2 g/dL |
| 23-666-388 | 1 x 15 mL | Total Protein Standard Set | 8 g/dL |
| 23-666-198 | 1 x 15 mL | Cholesterol Standard | 0.2 g/dL |
| 23-666-304 | 1 x 2 mL | Hemoglobin Standard | 12 g/dL |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Certifications/Compliance | CE |
| Color | Clear |
| Content And Storage | 2°C to 8°C |
| Concentration | 0.1 g/dL |
| Type | Glucose Standard |
| Includes | Sulfuric Acid (dilute Acid) 0.02%, Sodium Azide 0.01% |
| Quantity | 1 x 15 mL |
| Form | Liquid |
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?