Learn More
Thermo Scientific™ General Purpose Centrifuge Rotor Packages
GSA_VA
Catalog No. 75016106
Change view
Click to view available options
Model:
TX-1000 Blood Tube Package
TX-1000 Cell Culture Package
TX-1000 Microplate Package
TX-200 Blood Tube Package
TX-200 Cell Culture Package
TX-400 Blood Tube Package
TX-400 Cell Culture Package
TX-750 Blood Tube Package
TX-750 Cell Culture Package
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
75-016-106 TX-200 Cell Culture Package
75-016-103 TX-1000 Blood Tube Package
75-016-104 TX-1000 Microplate Package
75-016-105 TX-1000 Cell Culture Package
75-016-107 TX-400 Blood Tube Package
75-016-108 TX-400 Cell Culture Package
75-016-109 TX-750 Blood Tube Package
75-016-110 TX-750 Cell Culture Package
75-016-111 TX-200 Blood Tube Package
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 75-016-106 Supplier Thermo Scientific™ Supplier No. 75016106
Add to Cart
Add to Cart
Everything you need for your swing out rotor in one package
General Purpose Centrifuge Rotor packages with all the pieces you need to start spinning right away, including a rotor cross, bucket, lids and adapters
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Material See individual centrifuge, rotor and accessory data
Type Rotor Equipment Package
Capacity Max in TX-200: 20 x 15 mL Conical Tubes, 4 x 50 mL Conical Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
Net Weight See individual components of package
For Use With (Equipment) Purchase of Thermo Scientific General Purpose Centrifuge required for operation
Certifications/Compliance ISO 14971, ISO 13485, ISO 9001
Includes TX-200 with Buckets, ClickSeal Biocontainment lids, 15 and 50 mL conical tube adapters
Warranty See Manual
Biocontainment Yes
Model TX-200 Cell Culture Package