Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Value Lab Upright Freezers

Catalog No. 20LFEETSA
Change view
Click to view available options
Capacity (English):
1.8 cu. ft.
20.0 cu. ft.
3.5 cu. ft.
Electrical Requirements:
115 V 60 Hz
Type:
ADA Compliant Freezer
General Purpose Freezer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Electrical Requirements Type
20LFEETSA 20.0 cu. ft. 115 V 60 Hz General Purpose Freezer
02LFEETSA 1.8 cu. ft. 115 V 60 Hz General Purpose Freezer
04LFEETSA 3.5 cu. ft. 115 V 60 Hz General Purpose Freezer
04LFAETSA 3.5 cu. ft. 115 V 60 Hz ADA Compliant Freezer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 20LFEETSA Supplier Thermo Scientific™ Supplier No. 20LFEETSA

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Meet routine laboratory storage requirements with upright freezers that feature digital controls and excellent temperature stability.

Meet routine laboratory storage requirements with Thermo Scientific™ Value Lab Upright Freezers. Freezers feature digital controls and excellent temperature stability.

  • 20 cu. ft. upright
  • Four fixed wire shelves and six door shelves
  • Manual defrost
  • Enameled steel interior and exterior
  • CFC-free insulation and coolant

Warranty and Services

13 Months Parts and Labor

TRUSTED_SUSTAINABILITY

Specifications

Type General Purpose Freezer
Capacity (English) 20.0 cu. ft.
Capacity (Metric) 566.34 L
Temperature Range -12°C to -20°C
Temperature (Setpoint) Adjustable
Door Style Hinged
No. of Doors 1 (1 compartment)
Hinge Side Right
Insulation Polyurethane Foam
Interior Epoxy Coated Steel
Exterior Finish Epoxy Coated Steel
Shelves 4 Fixed
Door Shelves 6 Fixed
Certifications/Compliance cCSAus, cULus
Style Upright
Display Temperature Display - None
Casters Adjustable Leveling Feet
Color White
Control Type Mechanical Dial Thermostat (Inside Half Way Up Right Wall)
For Use With (Application) Indoor General Storage Of Non-Critical Samples And Reagents
Typical Lifetime 10 Years
Warranty Warranty varies by country, please contact us for details
Defrost Manual
Lighting Interior - Yes
Min. Clearance 3 in. On Sides, 3 in. At Back, 3 in. On Top
Model 20LFEETSA
Dimensions (D x W x H) Interior 23.8 x 27.8 x 57.8 in. (60.3 x 70.5 x 146.7 cm)
Dimensions (L x W x H) Exterior 32.13 x 34 x 73.13 in. (81.6 x 86.4 x 185.7 cm)
Amperage 5 A
Plug Type NEMA 5-15 P
Voltage 115 V
Electrical Requirements 115 V 60 Hz
Frequency 60 Hz
Wattage 575 w
Compressor Bottom-mounted
Refrigerant R134a, 100% CFC Free
Shipping Weight (English) 198 lb.
Shipping Weight (Metric) 89.8 kg
Rack Capacity 10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.