Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corepoint Scientific™ General Purpose Hazardous Location (Explosion Proof) Freezer
SDP

Catalog No. 22651301 Shop All Nor-Lake Scientific Products
Change view
Click to view available options
Capacity (Metric):
850 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22-651-301 850 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-651-301 Supplier Corepoint Scientific™ Supplier No. EF301WWW/0M

missing translation for 'validMainframeMsg'

This General Purpose Hazardous Location (Explosion Proof) Freezer is engineered for cold-storage use in environments where volatile conditions could potentially exist.

  • Meets NFPA and OSHA guidelines for hazardous locations
  • Mechanical thermostats sealed in UL listed explosion proof boxes
  • Keyed door locks
  • High density urethane foam cabinet and door insulation
  • Note: These units are not equipped with line cord and plug. They must be hardwired in conduit to a 120 VAC power source for safety.

Specifications

Type HazLoc Freezer
Capacity (English) 30 cu. ft.
Capacity (Metric) 850 L
Temperature Range -15°C to -25°C
Door Style Solid Swing
No. of Doors 1
Cabinet Material Scratch-resistant Painted Steel
Insulation Urethane
Interior Plastic
Exterior Finish Powder-coated Steel
Shelves 4 Fixed
Certifications/Compliance UL
Style Upright
Color White
Condenser Type Fan-forced
For Use With (Application) Frozen Substances
Warranty 1 year parts and labor warranty
Defrost Manual
Access Security Key Lock
Amperage 6 A
Plug Type No Plug, Hardwired
Voltage 100/115 V
Energy Usage 115 V, 60Hz, 1/3 HP
Compressor Top-mounted
Refrigerant CFC-free Refrigerant
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.