Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ General Purpose Tweezers, 12cm Long

Catalog No. 14955028 Shop All Fisherbrand Products
Change view
Click to view available options
Style:
With teeth
Without teeth
Description:
Blue
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style Description
14-955-028 With teeth White
14-955-029 Without teeth Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-955-028
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For precision handling without metal contact.

  • Made from polystyrene and high impact polystyrene (67% PS and 33% HIPS)
  • Disposable
  • Nonsterile
  • For handling parts and small samples
  • Available with or without tooth
  • Available in white and blue colors

Specifications

Length (Metric) 120 mm
Material Polystyrene and high impact polystyrene (67% PS and 33% HIPS)
Description White
For Use With (Application) Handling parts and small samples
Style With teeth
Product Type Tweezers, General Purpose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.