Learn More
3M™ General Use Masking Tape
Catalog No. 19070920 Shop All 3M Products
Change view
Click to view available options
Quantity:
24/Pk.
Catalog No. Quantity
19-070-920 24/Pk.
1 options
Catalog No. 19-070-920 Supplier 3M™ Supplier No. 7000144752
Add to Cart
Add to Cart
3M Company General Use Masking Tape is a proprietary, solvent-free adhesive that removes cleanly in one piece with no adhesive residue
  • Withstands surface temperatures up to 200°F (93°C) for 30 minutes
  • Sticks with confidence on wood, plastic, glass, metal and painted surfaces
  • Light tan backing tears easily and resists slivering
  • Low unwind force allows for rapid application and improved handling
  • Low unwind
  • 48 mm x 55 m 4.4 mil
  • 3M Catalog# 201+
TRUSTED_SUSTAINABILITY
Specifications
Color Tan
Material Crepe Paper
Product Type Masking and Duct Tapes
Model 201+
Quantity 24/Pk.