Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ GeneScan™ 500 ROX™ dye Size Standard

Catalog No. 4310361
Encompass_Preferred
Change view
Click to view available options
Quantity:
800 Reactions
9,600 injections
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-103-61 9,600 injections
40-173-4 800 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-103-61 Supplier Applied Biosystems™ Supplier No. 4310361
  • $11,640.00
    $10,592.70 / Each of 1

    Save $1,047.30

    9% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Includes

16 ROX™ dye-labeled, Single-stranded DNA fragments

The GeneScan™ 500 ROX™ Size Standard is an internal lane size standard developed for use with the Applied Biosystems fluorescence-based DNA electrophoresis systems.

The GeneScan™ 500 ROX™ Size Standard is an internal lane size standard developed for use with the Applied Biosystems fluorescence-based DNA electrophoresis systems. The use of an internal lane size standard enables automated data analysis and is also essential for achieving high run to run precision in sizing DNA fragments by electrophoresis. GeneScan™ 500 ROX™ Size Standard is designed for sizing DNA fragments in the 35-500 bp range and provides 16 single-stranded ROX™ dye labeled fragments of 35, 50, 75, 100, 139, 150, 160, 200, 250, 300, 340, 350, 400, 450, 490, and 500 bases. Each of the DNA fragments is labeled with a proprietary fluorophore, which results in a single peak when run under both denaturing and non-denaturing conditions.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Sequencing
Quantity 9,600 injections
Product Line GeneScan, ROX
For Use With (Equipment) 3730xl DNA Analyzer, 3730 DNA Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer
Label or Dye ROX
Product Type Dye
Run Time Standard
Content And Storage This kit contains ROX size standard and loading buffer.
Store the kit at 2°C to 8°C. (Do not freeze).
Description GeneScan™ 500 ROX™ dye Size Standard, ROX™ Label. Use with 3730xl DNA Analyzer, 3730 DNA Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.