Learn More
Ion Torrent™ Genexus™ Integrated Sequencer

Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Starting Material | DNA, RNA or cfTNA |
| Quantity | 1 sequencer |
| Product Line | Genexus |
| Shipping Condition | Room Temperature |
| Content And Storage | • Genexus Integrated Sequencer • Genexus Software • Power cord • Network cable |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.