Learn More
Scientific Industries Genie Incubate Shake W/Ratch 230V - UK Plug
Supplier: Scientific Industries SIG1503
GENIE Temp-Shaker 300 includes 2 Universal Ratcheting Clamps, provides high speed orbital shaking from 35 to 300 RPM. Precise temperature control from 28C to 75C, for use in many laboratory applications. The GENIE Temp-Shaker has a unique 3-way access clear dome chamber. Additional ratcheting clamps and other interchangable platforms styles and accessories available for purchase.
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.