Learn More
Enzo Life Sciences Genipin (25mg). CAS: 6902-77-8
SDP
Supplier:  Enzo Life Sciences ALX350383M025
Encompass_Preferred
Cell permeable inhibitor of uncoupling protein 2 (UCP2). Increases glucose-stimulated insulin secretion, mitochondrial membrane potential and ATP levels in pancreatic island cells. Induces apoptosis. Protein cross-linking agent. Anti-inflammatory and anti-angiogenic. Alternative name: (+)-Genipin, Methyl (1S,2R,6S)-2-hydroxy-9-(hydroxymethyl)-3-oxabicyclo[4.3.0]nona-4,8-diene-5-carboxylate. Purity: ≥98% (HPLC). Appearance: White to off-white crystalline solid. Solubility: Soluble in DMSO (50mg/ml), dimethyl formamide (25mg/ml), 100% ethanol (5mg/ml), methanol or ethyl acetate; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8873
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.