Learn More
TARGETMOL CHEMICALS INC GENKWANIN 20MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6S009520MG
Encompass_Preferred
Also available in 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Genkwanin (Apigenin 7-methyl ether) exerts its anti-inflammatory effect mainly through the regulation of the miR-11/MKP-1/MAPK pathway. 2. Genkwanin is transported by both passive diffusion and multidrug resistance protein (MDR)-mediated efflux mechanisms. purity: 98%
Catalog No. 50-257-6394