Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amsbio LLC (formerly Genlantis) Detachin™ Cell Detachment Reagent

Catalog No. p-3397797
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T100100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T100100 Supplier Amsbio LLC (formerly Genlantis) Supplier No. T100100

Gentle, rapid detachment of a wide range of cells

  • Maximum cell viability
  • Effective on a wide range of cells
  • No mammalian or bacterial byproducts
  • Stable at 4°C for two months
  • No need to wash detached cells
  • Suitable for detaching adherent cells from in vitro growth vesicles
  • Provides quick and gentle detachment and dissociation of a wide variety of adherent cells, including primary cells, grown in all known tissue culture plasticware

Specifications

Content And Storage -20°C
Product Type Cell Culture Dissociation Reagent
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.