Learn More
Amsbio LLC (formerly Genlantis) SoluLyse™ Bacterial Protein Extraction Reagent
Description
- Produces high yield of soluble proteins
- Simple one-step protocol
- Proprietary formulation of nonionic detergents optimized for bacterial protein extraction
- Proteins retain more activity
- Compatible with protease inhibitors, salts, chelating agents and reducing agents
- Choice of Phosphate or Tris buffer formulations to avoid interference with affinity purification resins and other downstream applications
Specifications
Specifications
| Formulation | 20mM Tris HCl Buffer, pH 7.5 |
| Quantity | 500 mL |
| Product Type | Bacterial Protein Extraction Reagent |
| For Use With (Application) | Proprietary formulation of nonionic detergents optimized for bacterial Protein extraction, high Protein yield with simple one-step proto col |
| Content And Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.