Learn More
ATCC Genomic DNA from Anaerococcus prevotii, Strain PC 1 - 5 μg (ATCC 9321D-5)
Supplier: ATCC 9321D5
Genomic DNA isolated from Anaerococcus prevotii (strain PC 1). Supplied as a 5 μg freeze-dried sample intended for research and bioinformatics applications; material is quality-verified and handled under BSL-1 conditions.
- Organism: Anaerococcus prevotii (strain PC 1).
- Product type: Genomic DNA.
- Format: Freeze-dried, 5 μg.
- Storage: 2-8°C.
- Applications: Genome sequencing and bioinformatics analyses.
- Biosafety level: BSL 1.
- Quality control: Verified by whole-genome sequencing.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.