Learn More
ATCC Genomic DNA from Bordetella pertussis strain 18323 - 5 μg (ATCC 9797D-5)
Supplier:  ATCC 18323
Encompass
Purified genomic DNA isolated from Bordetella pertussis strain 18323, supplied as a defined microgram quantity for molecular biology research applications such as PCR, sequencing, and assay development. Store refrigerated at 2-8 °C; intended for research use only.
  • Purified genomic DNA suitable for PCR and sequencing.
  • Derived from Bordetella pertussis strain 18323.
  • Supplied as 5 μg of DNA per vial.
  • Store at 2-8 °C; handle according to laboratory safety procedures.
  • Intended for research use only; not for diagnostic or therapeutic use.
  • Biosafety level 1 material.
Catalog No. NC3900621