Learn More
TRIBIOSCIENCE INC Genomic DNA Extraction from Cells and Tissues, 50 ML
SDP
Supplier:  TRIBIOSCIENCE INC TBS600850
Encompass_Preferred
Fast DNA extraction kit provides a simple and rapid method to isolate the genomic DNA from tissues or cells for PCR amplification, genotype identification, CRISPR mutation detection, and library screening.
Catalog No. NC2887467