Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HyClone™ Gentamicin Solution

Catalog No. SV3008001 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10 x 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SV3008001 10 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SV3008001 Supplier HyClone™ Supplier No. SV30080.01

This item is not available for purchase online. For pricing, please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

Control or destroy destructive microorganisms such as bacteria and mycoplasma. Cytiva HyClone™ Gentamicin solution is a broad spectrum cell culture antibiotic that is nontoxic to viruses and mammalian cells at antibacterial levels.

  • Active against gram positive and gram negative bacteria and mycoplasma; useful for long-term virus and tissue culture studies
  • Used as an antibacterial or antimycobacterial
  • Interferes with bacterial protein synthesis by binding to L6 protein of 50S ribosomal subunit
  • Common working concentration is 5μg/mL
  • Cytotoxic at >300μg/mL
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 3°C
Description Gentamicin Solution, 50mg/mL
Antibiotic Spectrum Broad Spectrum, Gram-Negative and Gram-Positive Bacteria
Concentration 50 mg/mL
Form Solution
Quantity 10 x 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.