Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentamicin sulfate solution-10, MP Biomedicals™

Catalog No. ICN10503010 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10503010 10 mL
ICN10503080 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10503010 Supplier MP Biomedicals, Inc Supplier No. 0210503010

Chemical Identifiers

CAS 1405-41-0
MDL Number MFCD00270181
Synonym Garamycin, Genoptic

Specifications

Type Gentamicin Sulfate
Physical Form Liquid
Color Colorless to Yellow
Quantity 10 mL
Grade USP
Chemical Name or Material Gentamycin Sulfate Solution - 10, Cell Biology and Analysis
EINECSNumber 215-778-9
Recommended Storage Store at 4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.