Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GeoSafari Bug Viewers
SCI_ED

Catalog No. S96066 Shop All Educational Insights Products
Change view
Click to view available options
3-inch
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S96066 3-inch
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96066 Supplier Educational Insights Supplier No. EI5111

missing translation for 'validMainframeMsg'

Get an extra close-up look at bugs of all sizes

Bug Viewer

  • Crystal clear plastic
  • High quality, 3 in. dia. magnifying lid
  • Built-in air holes and measuring grid
Jumbo Bug Viewer
  • Clear plastic container features a 4 in. dia. lid that kids can rotate for a close inspection
  • Extra “up close” inset magnifier
  • High-quality magnifying lid with built-in air holes and measuring grid on bottom of jar

Order Info

Grades K+

Specifications

Color Clear
Description Bug Viewer
Grade K
For Use With (Application) Viewing bugs of all sizes
Includes Magnifying lid, Built-in air holes, Measuring grid
Material Plastic
Size 3 in. dia.
Diameter (English) Outer 3 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.