Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Geranyl Tiglate 98.0+%, TCI America™
SDP

Catalog No. G043025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G043025ML 25 mL
G04305ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G043025ML Supplier TCI America Supplier No. G043025ML

Chemical Identifiers

CAS 7785-33-3
Molecular Formula C15H24O2
Molecular Weight (g/mol) 236.36
MDL Number MFCD00016622
InChI Key OGHBUHJLMHQMHS-KRDNBFTESA-N
Synonym Tiglic Acid Geranyl Ester
PubChem CID 5367785
IUPAC Name (2E)-3,7-dimethylocta-2,6-dien-1-yl (2E)-2-methylbut-2-enoate
SMILES C\C=C(/C)C(=O)OC\C=C(/C)CCC=C(C)C

Specifications

Color Yellow
Boiling Point 151°C
Quantity 25 mL
Formula Weight 236.36
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Geranyl Tiglate
EINECSNumber (2)-0970
RTECSNumber RG5927900
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.