Learn More
Echelon Biosciences Research Labs GERANYLS-THIOLODIPHOSPHATE 10 mg
SDP
Supplier:  Echelon Biosciences Research Labs IS10010MG
Encompass
Geranyl thiolodiphosphate (GSPP) is an analog of GPP in which the non-bridging oxygen has been replaced by sulfur resulting in a less-reactive compound. GSPP is turned over by avian farnesyl diphosphate synthase (FPPase) more slowly than FPP and acts as a competitive inhibitor (KI = 24.8 M). This item is non returnable
Catalog No. 50-720-413
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.