Learn More
TARGETMOL CHEMICALS INC GERMACRONE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2945
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Germacrone a major active component found in Curcuma zedoaria oil product shows anti-inflammatory antiandrogenic and antivirus activities. It also inhibits the proliferation of glioma cells by promoting apoptosis. purity: 99%
Catalog No. 50-257-7027
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.