Learn More
Neuvitro Gelatin coated sterile German glass coverslips for cell culture, 12mm dia #1 thick 90/pk, ready to use in cell culture.
Supplier: Neuvitro GG12GELATIN
CE certified German glass coverslips with Gelatin coating on both sides for cell culture and confocal / IHC imaging with no autofluorescence. Pre-washed, nitric acid treated, coated with porcine geltain and sterile. 12mm diameter round, #1 thickness, 90 pieces/pack. Ready to use directly in cell culture. Coverslips are sealed as 15 pieces per group for easy storage and enhanced cell culture quality. Shelf life 0.5 year when stored in chest freezer. User manual included.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.