Learn More
Neuvitro CE certified German glass coverslips for cell culture and confocal /IHC imaging, 12mm diameter #1.5 thick 0.5oz/pk untreated
Supplier:  Neuvitro GG121.5OZ
Encompass_Preferred
CE certified cell culture tested German glass coverslips for cell culture and confocal /IHC imaging, 12mm diameter #1.5 thickness 0.5oz/pk untreated. Pretreatment is required prior to cell culture applications. Shelf life 5 years, store at room temperature in dry.
Catalog No. NC1117320
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.