Learn More
Thermo Scientific Chemicals Gey's Balanced Salt Solution, with calcium, with magnesium, without phenol
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 500 mL |
Frequently Asked Questions (FAQs)
The Gey's Balanced Salt Solution, with calcium, with magnesium, without phenol contains:
- 121 mM sodium chloride
- 1.5 mM calcium chloride
- 4.9 mM potassium chloride
- 1.0 mM magnesium chloride
- 0.3 mM magnesium sulfate
- 27 mM sodium bicarbonate
- 0.2 mM monopotassium phosphate
- 0.8 mM sodium phosphate dibasic
- 5.6 mM glucose