Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ GFCI Extension Cable

Catalog No. 19104062 Shop All Brady Corporation Products
Change view
Click to view available options
GFCI extension cable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-104-062 GFCI extension cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-104-062 Supplier Brady™ Supplier No. ES14380

For outdoor use

  • Has highly visible yellow cable
  • Rated 15A/120V, 12/3 AWG

Specifications

Product Type Extension Cable
Length (English) 9 in.
Length (Metric) 22.86 cm
Color Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.