Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GFM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598790
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598790 Supplier Invitrogen™ Supplier No. PA598790
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GFM2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

GFM2 is involved in mitochondrial translation, which is crucial for maintaining mitochondrial function and mutations in this system lead to a breakdown in the respiratory chain-oxidative phosphorylation system and to impaired maintenance of mitochondrial DNA. In Eukaryotes contain two protein translational systems, one in the cytoplasm and one in the mitochondria. This gene encodes one of mitochondrial translation elongation factors. Its role in the regulation of normal mitochondrial function and in different disease states attributed to mitochondrial dysfunction is not known. Alternative splicing results in at least three transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFM2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Gfm2
Gene Accession No. Q969S9
Gene Alias 6530419G12Rik; A930009M04Rik; EFG2; EF-G2mt; EF-G2mt {ECO:0000255; Elongation factor G 2, mitochondrial; elongation factor G 2, mitochondrial {ECO:0000255; Elongation factor G2; G elongation factor; G elongation factor mitochondrial 2; G elongation factor, mitochondrial 2; GFM2; HAMAP-Rule:MF_03059}; hEFG2; mEF-G 2; mEF-G 2 {ECO:0000255; mitochondrial elongation factor G2; mitochondrial ribosome recycling factor 2; MRRF2; MST027; MSTP027; RGD1309854; ribosome-releasing factor 2, mitochondrial; ribosome-releasing factor 2, mitochondrial {ECO:0000255; RRF2; RRF2mt; RRF2mt {ECO:0000255
Gene Symbols Gfm2
Host Species Rabbit
Immunogen Recombinant Human Ribosome-releasing factor 2, mitochondrial protein (520-779AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84340
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.