Learn More
Cell Biolabs Inc GFP ELISA Kit, 96 assays
Supplier:  Cell Biolabs Inc AKR121
Encompass_Preferred
Although GFP expression can be easily detected under a fluorescence microscope, GFP fluorescence intensity varies from cell to cell because of the heterogeneity nature of GFP expression. In order to quantitate the GFP expression in cells, FACS analysis is usually required, which is both expensive and time consuming. Cell Biolabs’ GFP ELISA Kit is an enzyme immunoassay developed for detection and quantitation of GFP or GFP fusion protein in cell or tissue samples. The quantity of GFP or its variants (including BFP, CFP and YFP) in an unknown sample is determined by comparing its absorbance with that of a known recombinant GFP standard curve. The kit has detection sensitivity limit of 30 pg/mL GFP. The kit also provides an efficient system for rapid quantitation of GFP lentivirus titer for both viral supernatant and purified virus. Each kit provides sufficient reagents to perform up to 96 assays including standard curve and GFP samples.
Catalog No. NC0191128
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.