Learn More
Angio Proteomie cAP-0078GFP, GFP-Human Pancreatic Epithelioid Carcinoma Cells (PANC-1), 1 Frozen Vial, > 5x10[5] Cells/Vial
SDP
Supplier:  Angio Proteomie CAP0078GFP
Encompass
Human Pancreatic Epithelioid Carcinoma Cells (PANC-1) was derived from a Pancreatic Epithelioid Carcinoma of a 59-year old Caucasian male. GFP-Human Pancreatic Epithelioid Carcinoma Cells (PANC-1) are selected from the Human Pancreatic Epithelioid Carcinoma Cells (PANC-1) (cAP-0078) after transfected with GFP expressing lentiviruses resistant to puromycin. Cells are supplied in frozen vials with more than 5 x 10[5] cell/vial. Universal Full Growth Medium (cAP-01B) is recommended to culture the cells.
Catalog No. NC2736968
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.