Learn More
ABIOCODE INC GFP(M)ANTIBDY,MOUSE MONO,100UG
Supplier:  ABIOCODE INC M08024A
Encompass_Preferred
GFP (M) Antibody, Mouse MonoclonalQuantity: 100 ug Predicted I Observed M.W.: 27 kDa Isotype: Mouse IgG1
Catalog No. NC2204463
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.