Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Mouse anti-None with BSA, Clone: 3000000, MP Biomedicals™

Catalog No. MP08687402 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08687402 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP08687402 Supplier MP Biomedicals, Inc Supplier No. 08687402
Only null left
Add to Cart
Add to Cart

Mouse IgG2a monoclonal antibody (mAb) against GFP (clone 3E6) that recognizes the native form of the Green Fluorescent Protein (GFP), GFP variants and GFP-fusion proteins.

Anti-GFP, Clone 3E6, is a mouse IgG2a monoclonal antibody (mAb) that recognizes the native form of the Green Fluorescent Protein (GFP), GFP variants and GFP-fusion proteins. Used for the detection of GFP and variants in applications where the GFP is in a native form, such as immunoprecipitation, immunocytochemistry and ELISA analyses. Optimal dilutions must be determined by the researcher for each application.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFP
Applications Immunoassay, ELISA, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 3000000
Concentration 1.0 mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Clone 3E6
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species None
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.